Boc-L-Nle(6-OH) ≥99.5% (by HPLC), crystalline powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: (S)-2-((tert-Butoxycarbonyl)amino)-6-hydroxyhexanoic acid

047341GM
100277-366EA 311.23 USD
100277-366
Boc-L-Nle(6-OH) ≥99.5% (by HPLC), crystalline powder
Boc-L-Nle(6-OH)

  • White crystalline powder
  • Store at 0-8 °C

Formula: C₁₁H₂₁NO₅
MW: 247.3 g/mol
Melting Pt: 106…114 °C
MDL Number: MFCD01862292
CAS Number: 77611-37-1
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/64/39/21286439.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR