(S)-2-((tert-Butoxycarbonyl)amino)-4-phenylbutanoic acid ≥99.5%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: (S)-2-(Boc-amino)-4-phenylbutyric AcidN-Boc-L-homophenylalanineBoc-Homophe-OH(S)-2-(tert-Butoxycarbonylamino)-4-phenylbutyric Acid

047331GM
100281-714EA 85 USD
100281-714
(S)-2-((tert-Butoxycarbonyl)amino)-4-phenylbutanoic acid ≥99.5%
(S)-2-((tert-Butoxycarbonyl)amino)-4-phenylbutanoic acid
Formula: C₁₅H₂₁NO₄
MW: 279.34 g/mol
MDL Number: MFCD00076904
CAS Number: 100564-78-1
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/64/38/21286438.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR