Boc-Glu(OtBu)-OH ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

046835GM
100270-562EA 116.38 USD
100270-562
Boc-Glu(OtBu)-OH ≥99%
Boc-Glu(OtBu)-OH
Formula: C₁₄H₂₅NO₆
MDL Number: MFCD00038257
CAS Number: 13726-84-6
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/67/27/21286727.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR