Boc-D-Glu(OtBu)-OH ≥99%

Synonyms: N-Boc-D-glutamic acid 5-tert-butyl ester

046825GM
100297-856EA 73.03 USD
100297-856
Boc-D-Glu(OtBu)-OH ≥99%
Boc-D-Glu(OtBu)-OH
Formula: C₁₄H₂₅NO₆
MDL Number: MFCD00076927
CAS Number: 104719-63-3
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/75/05/21287505.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR