Boc-Dap(Fmoc)-OH ≥99%, crystalline powder

Supplier: INDOFINE CHEMICAL COMPANY MS

046491GM
100281-598EA 170 USD
100281-598
Boc-Dap(Fmoc)-OH ≥99%, crystalline powder
Boc-Dap(Fmoc)-OH

  • White crystalline powder
  • Store at 0-8 °C

Formula: C₂₃H₂₆N₂O₆
Melting Pt: 81…89 °C
MDL Number: MFCD00235897
CAS Number: 122235-70-5
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR