N-(tert-Butoxycarbonyl)-4-fluoro-D-phenylalanine ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: N-Boc-4-fluoro-D-phenylalanineBoc-D-Phe(4-F)-OH

046601GM
100274-262EA 116.38 USD
100274-262
N-(tert-Butoxycarbonyl)-4-fluoro-D-phenylalanine ≥99%
N-(tert-Butoxycarbonyl)-4-fluoro-D-phenylalanine
Formula: C₁₄H₁₈FNO₄
MW: 283.30 g/mol
MDL Number: MFCD00079669
CAS Number: 57292-45-2
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/61/76/21286176.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR