Boc-3,5-diiodo-L-tyrosine ≥99.5%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Boc-3,5-diiodo-Tyr-OH

046535GM
100281-608EA 251.08 USD
100281-608
Boc-3,5-diiodo-L-tyrosine ≥99.5%
Boc-3,5-diiodo-L-tyrosine
Formula: C₁₄H₁₇I₂NO₅
MW: 533.101 g/mol
MDL Number: MFCD00058515
CAS Number: 62129-53-7
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/67/20/21286720.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR