(S)-2-((tert-Butoxycarbonyl)amino)-2-cyclohexylacetic acid ≥99%, Powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: N-Boc-2-cyclohexyl-L-glycineBOC-L-α-cyclohexylglycineN-Boc-L-2-cyclohexylglycineBoc-Chg-OHBoc-cyclohexyl-Gly-OH

046205GM
100277-292EA 115.08 USD
100277-292
(S)-2-((tert-Butoxycarbonyl)amino)-2-cyclohexylacetic acid ≥99%, Powder
(S)-2-((tert-Butoxycarbonyl)amino)-2-cyclohexylacetic acid

  • White powder
  • Store at 0-8 °C

Formula: C₁₃H₂₃NO₄
MW: 257.3 g/mol
MDL Number: MFCD00133638
CAS Number: 109183-71-3
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/64/25/21286425.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR