N-(tert-Butoxycarbonyl)-4-chloro-D-phenylalanine ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: (R)-N-BOC-4-ChlorophenylalanineBoc-D-Phe(4-Cl)-OHN-Boc-4-chloro-D-phenylalanine

046151GM
100281-546EA 116.38 USD
100281-546
N-(tert-Butoxycarbonyl)-4-chloro-D-phenylalanine ≥99%
N-(tert-Butoxycarbonyl)-4-chloro-D-phenylalanine
Formula: C₁₄H₁₈ClNO₄
MW: 299.75 g/mol
MDL Number: MFCD00076978
CAS Number: 57292-44-1
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/67/12/21286712.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR