Boc-L-3-benzothienylalanine ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Boc-β-(3-benzothienyl)-Ala-OH(S)-3-(Benzo[b]thiophen-3-yl)-2-((tert-butoxycarbonyl)amino)propanoic acid

045871GM
100295-172EA 196.15 USD
100295-172
Boc-L-3-benzothienylalanine ≥99%
Boc-L-3-benzothienylalanine
Formula: C₁₆H₁₉NO₄S
MW: 321.391 g/mol
MDL Number: MFCD00237538
CAS Number: 154902-51-9
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/72/37/21287237.jpg https://us.vwr.com/stibo/web/std.lang.all/72/36/21287236.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR