N-Boc-L-aspartic acid-4-tert-butyl ester dicyclohexylammonium salt ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: N-α-t-Boc-L-Aspartic Acid β-t-butyl Ester Dicyclohexylammonium SaltBoc-Asp(OtBu)-OH·DCHA

045805GM
100281-490EA 122.92 USD
100281-490
N-Boc-L-aspartic acid-4-tert-butyl ester dicyclohexylammonium salt ≥99%
N-Boc-L-aspartic acid-4-tert-butyl ester dicyclohexylammonium salt
Formula: C₁₃H₂₃NO₆·C₁₂H₂₃N
MW: 470.651 g/mol
MDL Number: MFCD00038889
CAS Number: 1913-12-8
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/67/03/21286703.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR