Boc-Asp-(OBzl)-Phe-OH ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

045791GM
100281-488EA 285.08 USD
100281-488
Boc-Asp-(OBzl)-Phe-OH ≥99%
Boc-Asp-(OBzl)-Phe-OH
Formula: C₂₅H₃₀N₂O₇
MDL Number: MFCD00270492
CAS Number: 68763-45-1
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/64/16/21286416.jpg https://us.vwr.com/stibo/web/std.lang.all/64/17/21286417.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR