Boc-ARG(Boc)2-OH ≥97% (by HPLC), powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Boc-Arg(Boc)₂-OH

045571GM
100292-890EA 496.92 USD
100292-890
Boc-ARG(Boc)2-OH ≥97% (by HPLC), powder
Boc-ARG(Boc)2-OH

  • White powder
  • Store at 0-8 °C

Formula: C₂₁H₃₈N₄O₈
MW: 474.555 g/mol
Melting Pt: 116…120 °C
MDL Number: MFCD00236819
CAS Number: 97745-69-2
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR