1-Boc-4-(aminomethyl)piperidine ≥95%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: tert-Butyl 4-(aminomethyl)piperidine-1-carboxylatetert-Butyl 4-(aminomethyl)tetrahydropyridine-1(2H)- carboxylate4-(Aminomethyl)-1-tert-butoxycarbonylpiperidine4-(Aminomethyl)-1-Boc-piperidine4-(Aminomethyl)-1-piperidinecarboxylic acid tert-Butyl ester1-Boc-4-(Aminomethyl)piperidinetert-Butyl 4-(Aminomethyl)-1-piperidinecarboxylate

045461GM
100295-142EA 31.38 USD
100295-142
1-Boc-4-(aminomethyl)piperidine ≥95%
1-Boc-4-(aminomethyl)piperidine

  • Light yellow oil or low melting point crystalline mass
  • Store at 0-8 °C

Formula: C₁₁H₂₂N₂O₂
MW: 214.31 g/mol
MDL Number: MFCD01076207
CAS Number: 144222-22-0
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/73/84/21287384.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR