Mono(boc)(PEO)-3-diamine, boc-DOOA ≥98% (by TLC)

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: tert-Butyl [2-[2-(2-Aminoethoxy)ethoxy]ethyl]carbamate[2-[2-(2-Aminoethoxy)ethoxy]ethyl]carbamic acid tert-Butyl esterN-(tert-Butoxycarbonyl)-2,2'-(ethylenedioxy)diethylamineN-Boc-2,2'-(ethylenedioxy)diethylamineN-(tert-Butoxycarbonyl)-3,6-dioxa-1,8-octanediamineN-Boc-3,6-dioxa-1,8-octanediamine

045411GM
100295-140EA 338.69 USD
100295-140
Mono(boc)(PEO)-3-diamine, boc-DOOA ≥98% (by TLC)
Mono(boc)(PEO)-3-diamine, boc-DOOA

  • Yellow oil
  • Store at 0-8 °C

Formula: C₁₁H₂₄N₂O₄
MW: 248.3 g/mol
MDL Number: MFCD03788155
CAS Number: 153086-78-3
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/73/83/21287383.jpg https://us.vwr.com/stibo/web/std.lang.all/73/82/21287382.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR