1-(Boc-amino)cyclopropanecarboxylic acid ≥99% (by HPLC), crystalline powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 1-Aminocyclopropane-1-carboxylic acid, N-BOC protected1-(Boc-Amino)cyclopropanecarboxylic acid

045401GM
100281-456EA 119 USD
100281-456
1-(Boc-amino)cyclopropanecarboxylic acid ≥99% (by HPLC), crystalline powder
1-(Boc-amino)cyclopropanecarboxylic acid

  • White crystalline powder
  • Store at 0-8 °C

Formula: C₉H₁₅NO₄
MW: 201.22 g/mol
Melting Pt: 175 °C
MDL Number: MFCD00083257
CAS Number: 88950-64-5
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/64/13/21286413.jpg https://us.vwr.com/stibo/web/std.lang.all/64/14/21286414.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR