Boc-Abu-OH ≥99%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Boc-L-2-aminobutanoic acidBOC-L-α-Aminobutyric acid(S)-2-(tert-Butoxycarbonylamino)butyric acidBoc-L-Abu-OH(S)-2-(Boc-amino)butyric acid

045205GM
100271-586EA 156.92 USD
100271-586
Boc-Abu-OH ≥99%, powder
Boc-Abu-OH

  • White to off-white powder
  • Store at 0-8 °C

Formula: C₉H₁₇NO₄
MW: 203.238 g/mol
MDL Number: MFCD00037267
CAS Number: 34306-42-8
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR