Boc-D-Dap(Aloc)-OH DCHA

Supplier: INDOFINE CHEMICAL COMPANY MS

046331GM
100277-302EA 311.23 USD
100277-302
Boc-D-Dap(Aloc)-OH DCHA
Boc-D-Dap(Aloc)-OH DCHA
Formula: C₁₂H₂₁N₂O₆·C₁₂H₂₃N
MDL Number: MFCD00800347
CAS Number: 204197-26-2
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/67/16/21286716.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR