11-(4,4-difluoro-5,7-dimethyl-4-bora-3a,4a-diaza-s-indacene-3-yl)Undecanoic Acid NHS Ester, Marker Gene Technologies, Inc.
M1530
101743-068EA 433.38 USD
101743-068
11-(4,4-difluoro-5,7-dimethyl-4-bora-3a,4a-diaza-s-indacene-3-yl)Undecanoic Acid NHS Ester, Marker Gene Technologies, Inc.
BODIPY® FL C11, SE equivalent
  • Amine-reactive probe
  • Substitute for fluorescein
  • Provides pH-insensitive fluorescence
This fluorescent amine-reactive probe has an added spacer to improve accessibility of the reactive group by nucleophiles in preparation of bioconjugates and can be used as a substitute for fluorescein providing pH-insensitive fluorescence and narrow spectral bandwidth.