SP Wilmad-LabGlass General Purpose Distilling Apparatus, SP Industries
Supplier: SP Industries
LG-6600-120 LG-6600-122 LG-6600-130 LG-6600-102
14204-862EA 257.5 USD
14204-862 14203-356 14208-828 14206-018
SP Wilmad-LabGlass General Purpose Distilling Apparatus, SP Industries
These borosilicate glass apparatus are used in general purpose and phenol distillation
They are useful for batch production of high purity water. Complete unit consists of a flask with a flat bottom and distillation head having Standard Taper 19/38 joint, Standard Taper Stopper, and Graham style 200mm long jacketed condenser with a Standard Taper 19/38 joint at top only.