Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Taq DNA Polymerase-With XD Buffer, MP Biomedicals, LLC

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
11EPTQX100 11EPTQX025 11EPTQX925
Taq DNA polymerase is now available with an incubation buffer containing no detergent and no BSA.

  • Highly reproducible results
  • Ideal for T/A cloning
  • Ultra pure enzyme
  • Highly processive

Taq DNA Polymerase is a specialized thermostable enzyme isolated from the thermophilic bacterium Thermus aquaticus

Taq polymerase catalyzes oligonucleotide primer-driven, DNA template dependent incorporation of dNTPs into complimentary DNA strands. It displays both 5′ to 3′ polymerase and exonuclease activities.

One unit will incorporate 10 nmol of total dNTPs into acid-precipitable DNA in 30 min at 74° C.

The recombinant form of this enzyme is expressed in E. coli. Taq DNA polymerase is now available with an incubation buffer containing no detergent and no BSA. This incubation buffer (XD buffer) allows optimal results for researchers working with vegetal or beef material or using further dHPLC.




 

ORDER

Expand All / Collapse All
Description Size Supplier No. VWR Catalog Number Unit Price Quantity
DNA Polymerase-With XD Buffer 250 Units 11EPTQX025 75857-040 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

DNA Polymerase-With XD Buffer 1000 Units 11EPTQX100 75856-996 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

DNA Polymerase-With XD Buffer 10 X 250 Units 11EPTQX925 75856-998 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Application Nucleic Acid Amplification, PCR, RT-PCR, DNA sequencing and DNA labeling
Concentration 5U/ųL
Storage Temperature Store at -20° C
Buffer Composition 10X XD Buffer with MgCl2 Incubation buffer XD with MgCl2 (1x): 20mM Tris-HCl pH8.3, 50mM Kcl, 1.5mM MgCl2
Product Type Proteins, Enzymes & Peptides
Format Liquid
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping