Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Taq CORE Kit 25, MP Biomedicals, LLC

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
11EPTQK251 11EPTQK255C 11EPTQK253 11EPTQK25L
Taq DNA Polymerase + dNTPs in one kit.

  • Contains dNTPs at 25mM
  • Highly reproducible results
  • Ideal for T/A cloning
  • Ultra pure enzyme
  • Highly processive

Taq CORE Kits contain all the reagents (presented as separate items) required for PCR: MP Biomedicals high quality recombinant Taq DNA polymerase, high purity dNTPs Mix (>99%), optimized incubation buffers with and without MgCl2 and a separate MgCl2 solution.

Highly processive, recombinant, thermostable 5'-3' DNA polymerase catalyzes the addition of mononucleotide units to the 3'-OH end of a primer chain. Remains functional even after prolonged incubation at 95°C. This enzyme possesses 5'-3' exonucleases activity.

Quality control performed on every lot of enzymes: Specific PCR on genomic and phage templates, activity check, absence of ribonucleases, absence of nickases and endonucleases, absence of 3' exonucleases, purity on SDS-PAGE, absence of proteases.

Unit Definition: One unit is the amount of enzyme required to catalyze the incorporation of 10nmol of nucleotides into acid-insoluble material in 30min at 74°C under assay conditions.




 

ORDER

Expand All / Collapse All
Description Size Supplier No. VWR Catalog Number Unit Price Quantity
CORE Kit 25 250 Units 11EPTQK251 75857-026 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

CORE Kit 25 3 X 250 Units 11EPTQK253 75857-028 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

CORE Kit 25 5 X 250 Units 11EPTQK255C 75857-030 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

CORE Kit 25 5 X 1000 Units 11EPTQK25L 75857-032 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Application Nucleic Acid Amplification, PCR, RT-PCR, DNA sequencing and DNA labeling
Concentration 5U/ųL
Storage Temperature Store at -20° C
Buffer Composition Standard buffer w/MgCl2 Standard buffer w/o MgCl2 25mM MgCl2 solution
Components PCR reagents, Taq DNA polymerase, high purity dNTPs at 25mM each Mix (99%), optimized incubation buffers with and without MgCl2 and a separate MgCl2 solution.
Product Type Kits & Assays
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping