Taq CORE Kit 10, MP Biomedicals

Supplier: MP Biomedicals
11EPTQK105C 11EPTQK10L 11EPTQK101 11EPTQK103 11EPTQK103 11EPTQK300 11EPTQK109 11EPTQK300 11EPTQK101 11EPTQS901K 11EPTQK10L 11EPTQK105C 11EPTQK109
75857-020EA 376.04 USD
75857-020 75857-022 76020-292 CA76046-318 CA76046-254 CA76046-256 CA76045-946 75857-018 76020-296 CA76046-250 75857-024 76020-302 CA76046-252
Taq CORE Kit 10, MP Biomedicals
Taq DNA Polymerase + dNTPs in one kit.

  • Contains dNTPs at 10mM
  • Highly reproducible results
  • Ideal for T/A cloning
  • Ultra pure enzyme
  • Highly processive

Taq CORE Kits contain all the reagents (presented as separate items) required for PCR: MP Biomedicals high quality recombinant Taq DNA polymerase, high purity dNTPs Mix (>99%), optimized incubation buffers with and without MgCl2 and a separate MgCl2 solution.
Applications: Nucleic Acid Amplification, PCR, RT-PCR, DNA sequencing and DNA labeling

Highly processive, recombinant, thermostable 5'-3' DNA polymerase catalyzes the addition of mononucleotide units to the 3'-OH end of a primer chain. Remains functional even after prolonged incubation at 95°C. This enzyme possesses 5'-3' exonucleases activity.

Quality control performed on every lot of enzymes: Specific PCR on genomic and phage templates, activity check, absence of ribonucleases, absence of nickases and endonucleases, absence of 3' exonucleases, purity on SDS-PAGE, absence of proteases.

Unit Definition: One unit is the amount of enzyme required to catalyze the incorporation of 10nmol of nucleotides into acid-insoluble material in 30min at 74°C under assay conditions.

Components: PCR reagents, Taq DNA polymerase, high purity dNTPs at 10mM each Mix (>99%), optimized incubation buffers with and without MgCl2 and a separate MgCl2 solution.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR