Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Taq DNA Polymerase, MP Biomedicals, LLC

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
11EPTQD025 11EPTQD925 11EPTQD325
Highly processive, recombinant, thermostable 5'-3' DNA polymerase with 10X Buffer with MgCl2 only.

  • Highly reproducible results
  • Ideal for T/A cloning
  • Ultra pure enzyme
  • Highly processive

Taq DNA Polymerase is a specialized thermostable enzyme isolated from the thermophilic bacterium Thermus aquaticus

Taq polymerase catalyzes oligonucleotide primer-driven, DNA template dependent incorporation of dNTPs into complimentary DNA strands. It displays both 5′ to 3′ polymerase and exonuclease activities.

Quality control performed on every lot of enzymes: Specific PCR on genomic and phage templates, activity check, absence of ribonucleases, absence of nickases and endonucleases, absence of 3' exonucleases, purity on SDS-PAGE, absence of proteases.

One unit will incorporate 10 nmol of total dNTPs into acid-precipitable DNA in 30 min at 74 °C.




 

ORDER

Expand All / Collapse All
Description Size Supplier No. VWR Catalog Number Unit Price Quantity
DNA Polymerase 250 Units 11EPTQD025 75857-012 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

DNA Polymerase 3 x 250 Units 11EPTQD325 75857-014 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

DNA Polymerase 10 x 250 Units 11EPTQD925 75857-016 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Application Nucleic Acid Amplification, PCR, RT-PCR, DNA sequencing and DNA labeling
Concentration 5U/ųL
Storage Temperature Store at -20° C
Buffer Composition 10X Buffer with MgCl2
Components 20mM Tris-HCL pH 8.0, 100mM KCl, 0.1 mM EDTA, 1mM dithiothreitol, 50% glycerol, 10X buffer with MgCl4 20mM Tris-HCL pH 8.0, 100mM KCl, 0.1 mM EDTA, 1mM dithiothreitol, 50% glycerol, 10X buffer with MgCl3 20mM Tris-HCL pH 8.0, 100mM KCl, 0.1 mM EDTA, 1mM dithiothreitol, 50% glycerol, 10X buffer with MgCl2
Product Type Proteins, Enzymes & Peptides
Format Liquid
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping