Taq DNA Polymerase, Concentrated, MP Biomedicals
Supplier: MP Biomedicals
11EPTQC060 11EPTQC200
75856-994EA 899.58 USD
75856-994 75856-992
Taq DNA Polymerase, Concentrated, MP Biomedicals
Avoid inadvertently raising glycerol concentrations.
  • Highly reproducible results
  • Ideal for TA cloning
  • Ultra pure enzyme
  • Highly processive
Certain PCR applications require a high concentration of Taq DNA Polymerase
Highly processive, recombinant, thermostable 5'-3' DNA polymerase catalyzes the addition of mononucleotide units to the 3'-OH end of a primer chain. Remains functional even after prolonged incubation at 95° C. This enzyme possesses 5'-3' exonucleases activity.
Quality control performed on every lot of enzymes: Specific PCR on genomic and phage templates, activity check, absence of ribonucleases, absence of nickases and endonucleases, absence of 3' exonucleases, purity on SDS-PAGE, absence of proteases.
Unit Definition: One unit is the amount of enzyme required to catalyze the incorporation of 10nmol of nucleotides into acid-insoluble material in 30 minutes at 74° C under assay conditions.