Taq DNA Polymerase, MP Biomedicals

Supplier: MP Biomedicals
11EPTQA325 11EPTQA025 11EPTQA105 11EPTQA500 11EPTQA100 11EPTQA925
75856-990EA 1043.15 USD
75856-990 75857-142 75856-988 75856-986 75856-984 75856-982
Taq DNA Polymerase, MP Biomedicals
Highly processive, recombinant, thermostable 5'-3' DNA polymerase.

  • Highly reproducible results
  • Ideal for T/A cloning
  • Ultra pure enzyme
  • Highly processive

DNA polymerase catalyzes the addition of mononucleotide units to the 3'-OH end of a primer chain

Quality control performed on every lot of enzymes: Specific PCR on genomic and phage templates, activity check, absence of ribonucleases, absence of nickases and endonucleases, absence of 3' exonucleases, purity on SDS-PAGE, absence of proteases.

Unit Definition: One unit is the amount of enzyme required to catalyze the incorporation of 10nmol of nucleotides into acid-insoluble material in 30 min at 74 °C under assay conditions.

Remains functional even after prolonged incubation at 95°C. This enzyme possesses 5'-3' exonucleases activity.

Components: 20mM Tris-HCL pH 8.0, 100mM KCl, 0.1 mM EDTA, 1mM dithiothreitol, 50% glycerol, Incubation buffer with MgCl₂, Incubation buffer without MgCl₂
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR