    Sort by:
  • Search Within Results
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Marker Gene Technologies
Description:   Highly polar, highly water soluble lipase substrate is a derivative of HPTS (pyranine) that forms tight micelles in aqueous solutions.