    Sort by:
  • Search Within Results
Dansyl-11-aminoundecanoic acid
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Catalog Number: (101447-536)
Supplier:  Marker Gene Technologies
Description:   Ambiphilic probe with a polar, environmentally-sensitive dansyl fluorophore attached to a long chain fatty acid. The polar dansyl headgroup partitions...