    Sort by:
  • Search Within Results
5(6)-Carboxynaphthofluorescein
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Catalog Number: (101447-535)
Supplier:  Marker Gene Technologies
Description:   This pH-dependent fluorophore with red fluorescence (EX 598/EM 668 nm at pH 9) has a high pKa (7.6) value. The fluorescence shifts to shorter waveleng...