    Sort by:
  • Search Within Results
5-Fluorouridine-5'-O-β-D-galactopyranoside
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Marker Gene Technologies
Description:   Analog of 5-fluorouridine (5-FUR) useful for lacZ specific release of 5-FUR in recombinant cells or tissues.