You Searched For: Biocytin hydrazide hydrochloride
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Catalog Number: (76481-652)
Supplier: AAT BIOQUEST INC
Description: This water-soluble biocytin hydrazide is used for the selective non-radioactive detection of glycoconjugates.
Minority or Woman-Owned Business Enterprise Small Business Enterprise