    Sort by:
  • Search Within Results
Fluorescein di-β-D-glucopyranoside
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Catalog Number: (101176-338)
Supplier:  Marker Gene Technologies
Description:   Ultrasensitive fluorogenic substrate for quantitative measurement of β-glucosidase activity in vivo or in vitro.