    Sort by:
  • Search Within Results
You Searched for: "myo-inositol-galactoside"
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Catalog Number: (101176-252)
Supplier:  Marker Gene Technologies
Description:   This galactoside derivative of myo-inositol will release myo-inositol upon action of β-galactosidase.