Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Curriculum Mastery® Review Cards

Supplier: NEWPATH LEARNING LLC
446007 446011 446012 449007
Provides for an Engaging Review

  • 750 illustrated review cards
  • Covers a span of 25 key topics with 30 questions each

This set ensures that students obtain both the most expansive and comprehensive coverage of biology topics. Given in a fun and engaging manner that is easy to implement, students will learn about these standards based topics including bacteria and viruses, cells, and genetics just to name a few. Each study card set is accompanied by a Classroom Presentation/Assessment CD that features review questions given in an interactive flashcard mode. Also included is a Resource/Instruction card that offers suggestions based on whether this will be implemented in an individual manner or with a group setting. Use this on an interactive whiteboard or a computer for both reinforcement and review.



 

ORDER

Expand All / Collapse All
Description Supplier No. VWR Catalog Number Unit Price Quantity
Biology Flash Cards, 750 Cards 449007 470144-554 Case of 8 Retrieving
Each Retrieving
Human Body Flash Cards, 750 Cards 446011 470144-718 Case of 8 Retrieving
Each Retrieving
Life Science Flash Cards, 750 Cards 446007 470144-484 Case of 8 Retrieving
Each Retrieving
Six Kingdoms Flash Cards, 750 Cards 446012 470144-638 Case of 8 Retrieving
Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Length 7 cm
Width 5 cm

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty