GSK1070916 >97%

Supplier: SELLECK CHEMICALS LLC MS

S2740-50MG S2740-10MG S2740-5MG
101759-288EA 273.85 USD
101759-288 101759-290 101759-292
GSK1070916 >97%
GSK1070916

GSK1070916 is a reversible and ATP-competitive inhibitor of Aurora B/C with IC50 of 3.5 nM/6.5 nM


It displays >100-fold selectivity against the closely related Aurora A-TPX2 complex.

Storage Temperature: Freezer
CAS Number: 942918-07-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR