VetScan® i-STAT®1 Analyzer and Cartridges, Abaxis
Supplier: Abaxis
Vetscan
600-9004-25 10023272 10023275 600-9007-25
89126-038PK 975.84 USD
89126-038 89126-036 89126-044 89126-050
VetScan® i-STAT®1 Analyzer and Cartridges, Abaxis
The VetScan i-STAT® 1 delivers accurate blood gas, electrolyte, chemistry and hematology results in minutes from 2 to 3 drops of whole blood.
  • Small sample size 95 μl or less
  • Accurate and reliable
  • Portable, lightweight, weighs about 1.4 lbs.
  • Integrated barcode reader
  • Cardiac Troponin I Cartridge
Handheld package—making it the ideal solution for critical care situations, hospital operating room monitoring, exotic animals and research needs at the point of care.
The VetScan i-STAT 1 provides the additional information needed to monitor chronic disease patients, evaluate pre-anesthetic patients and provide diagnostic specialty testing information for the ill patient in the exam or treatment room.
The VetScan i-STAT 1 uses a wide range of disposable, single-use cartridges that contain the necessary reagents to provide reference lab quality results, while improving efficiency throughout the animal health continuum of care.