VWR® Alcohol Prep Pad, Sterile
Supplier: Avantor
PHX1007
75856-902BX 3.93 USD
75856-902
VWR® Alcohol Prep Pad, Sterile
Sterile, two-ply, folded, spun lace, alcohol prep pads are saturated with 70% isopropyl alcohol (Antiseptic).
  • Soft and absorbent, non-woven pads
  • Latex-free
The soft and absorbent pads are gamma sterilized.
They weigh 40 g per square meter (GSM).
Certifications: Manufactured according to FDA, GMP, and USP standards.