COULTER CLENZ® Cleaning Agent, Beckman Coulter
Supplier: Beckman Coulter
8546929 8546930 8546931
BK8546929EA 75.7 USD
BK8546929 BK8546930 BK8546931
COULTER CLENZ® Cleaning Agent, Beckman Coulter
Cleaning Agents Decontaminants
COULTER CLENZ® Cleaning Agent that strip away protein build-up and lysed red blood cells in instrument fluidics.
  • Strips away proteins
  • Aids preventative maintenance
It should be included in a schedule of routine preventive maintenance procedures designed to maintain high performance.