FastPrep™ FastDNA™-96 Fecal DNA Kit, MP Biomedicals, LLC
Supplier: MP Biomedicals
FastPrep™ FastDNA™
119696400
75856-394EA 1268.78 USD
75856-394
FastPrep™ FastDNA™-96 Fecal DNA Kit, MP Biomedicals, LLC
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The FastDNA-96™ Fecal DNA Kit quickly and efficiently isolates inhibitor-free, PCR-ready genomic DNA from mammalian stool samples Mammalian Stool (i.e. humans, primates, birds, rodents, cattle, canine, feline, porcine, etc.).
  • Lyse a wide variety of eukaryotic and prokaryotic organisms
  • Specially formulated lysis solution
  • Eluted DNA is ready for downstream applications
  • Procedure can be performed in minutes
  • No need for organic denaturants or proteinases
The FastDNA™-96 series of kits are designed for the simple and rapid isolation of high purity, PCR-ready DNA from human, animal and plant tissues, microbes, soil and other samples
The purification step removes all PCR inhibitors commonly found in a variety of dirty and difficult samples such as wastewater, fecal materials, soils, sediments and others. Samples are rapidly ground, homogenized and lysed. Cells are efficiently lysed by bead beating insided FastPrep™-96 system.
FastSpin plate and other standard format technology is then used to isolate the DNA, which, if necessary, is subsequently filtered to remove humic acids and polyphenols which can inhibit PCR. The procedure can be performed in minutes. There is no need for organic denaturants or proteinases.
This series of products can be used to isolate DNA from tough-to-lyse samples as well as from dirty (highly contaminated) samples such as forensic samples, bacteria, fungi, protozoa and algae.
Packaging: FastDNA™-96 Lysing Matrix Rack (0.5 mm Beads) 2 x 96-well rack, Lysis Buffer (2 x 40 mL), Fecal DNA Binding Solution (150 mL), Binding Plate Pre-Wash Buffer (50 mL), Fecal DNA Wash Buffer (100 mL), Elution Solution (2 x 10 mL), Inhibitor Plate Prep Solution (30 mL), Deep-Well Plate (2 each), MP-96 Binding Plate (2 each), MP-96 Inhibitor Removal Plate (2 each), Collection Plate (2 each), Elution Plate (6 each), Foil Plate Cover (4 each).
Caution: Store at Room temperature (15-30 °C)