FastPrep™ FastDNA™-96 Fungal/Bacterial Kit, MP Biomedicals, LLC

Supplier: MP Biomedicals
FastPrep™ FastDNA™
119696300
75856-392EA 1268.78 USD
75856-392
FastPrep™ FastDNA™-96 Fungal/Bacterial Kit, MP Biomedicals, LLC
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
FastDNA™-96 Fungal/Bacterial DNA Kit is used for rapid isolation of PCR-quality DNA from tough-to-lyse fungi and bacteria in approximately 40 minutes.

  • Procedure can be completed in minutes
  • Samples are rapidly and efficiently lysed by bead beating
  • Isolated DNA is ideal for downstream molecular-based applications
  • Sample type: Bacteria (Gram +/-), Yeast, Fungi, Spores, Algae

The FastDNA™-96 series of kits are designed for the simple and rapid isolation of high purity, PCR-ready DNA from human, animal and plant tissues, microbes, soil and other samples

The purification step removes all PCR inhibitors commonly found in a variety of dirty and difficult samples such as wastewater, fecal materials, soils, sediments and others. Samples are rapidly ground, homogenized and lysed. Cells are efficiently lysed by bead beating insided FastPrep™-96 system.

FastSpin plate and other standard format technology is then used to isolate the DNA, which, if necessary, is subsequently filtered to remove humic acids and polyphenols which can inhibit PCR. The procedure can be performed in minutes. There is no need for organic denaturants or proteinases.

This series of products can be used to isolate DNA from tough-to-lyse samples as well as from dirty (highly contaminated) samples such as forensic samples, bacteria, fungi, protozoa and algae.

Store at Room temperature (15-30 °C)

Packaging: Components: FastDNA-96™ Lysing Matrix Rack (0.5 mm Beads) 2 x 96-well rack, Lysis Buffer (2 x 40 mL), Fungal/Bacterial DNA Binding Solution (150 mL), Binding Plate Pre-Wash Buffer (50 mL), Fungal/Bacterial DNA Wash Buffer (100 mL), Elution Solution (2 x 10 mL), Deep-Well Plate (2 each), MP-96 Binding Plate (2 each), Collection Plate (2 each), Elution Plate (2 each), Foil Plate Cover (4 each).
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR