Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

LiChrosorb® NH2, ES Industries

Supplier: ES INDUSTRIES
LiChrosorb®
135221LNH2 134221LNH2 165321LNH2 164321LNH2 165221LNH2 155321LNH2 154321LNH2 155221LNH2 164221LNH2 144321LNH2 145321LNH2 145221LNH2 154221LNH2 144221LNH2
LiChrosorb is a high quality irregular silica with a high surface area and excellent loadability.




 

ORDER

Expand All / Collapse All
Column Format Phase I.D. x L Particle Size Pore Size Supplier No. VWR Catalog Number Unit Price Quantity
Analytical NH2 4.0x150 mm 5 µm 100 Å 134221LNH2 101308-538 Each Retrieving
View MSDS SDS
Analytical NH2 4.0x200 mm 5 µm 100 Å 144221LNH2 101308-542 Each Retrieving
View MSDS SDS
Analytical NH2 4.0x250 mm 5 µm 100 Å 154221LNH2 101308-546 Each Retrieving
View MSDS SDS
Analytical NH2 4.0x300 mm 5 µm 100 Å 164221LNH2 101308-550 Each Retrieving
View MSDS SDS
Analytical NH2 4.6x150 mm 5 µm 100 Å 135221LNH2 101308-540 Each Retrieving
View MSDS SDS
Analytical NH2 4.6x200 mm 5 µm 100 Å 145221LNH2 101308-544 Each Retrieving
View MSDS SDS
Analytical NH2 4.6x250 mm 5 µm 100 Å 155221LNH2 101308-548 Each Retrieving
View MSDS SDS
Analytical NH2 4.6x300 mm 5 µm 100 Å 165221LNH2 101308-552 Each Retrieving
View MSDS SDS
Analytical NH2 4.0x200 mm 10 µm 100 Å 144321LNH2 101308-526 Each Retrieving
View MSDS SDS
Analytical NH2 4.0x250 mm 10 µm 100 Å 154321LNH2 101308-530 Each Retrieving
View MSDS SDS
Analytical NH2 4.0x300 mm 10 µm 100 Å 164321LNH2 101308-534 Each Retrieving
View MSDS SDS
Analytical NH2 4.6x200 mm 10 µm 100 Å 145321LNH2 101308-528 Each Retrieving
View MSDS SDS
Analytical NH2 4.6x250 mm 10 µm 100 Å 155321LNH2 101308-532 Each Retrieving
View MSDS SDS
Analytical NH2 4.6x300 mm 10 µm 100 Å 165321LNH2 101308-536 Each Retrieving
View MSDS SDS
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty