Would you like to log in again?
Pseudalert Test, Idexx
Supplier: IDEXX DISTRIBUTION CORP.
Pseudalert Test detects the presence of Pseudomonas aeruginosa in water samples.
  • Less than 1 minute of hands-on time for streamlined testing work flow
  • Blue fluorescence of wells or vessel indicates a positive result
Pseudalert Test is based on a bacterial enzyme detection method that signals the presence of Pseudomonas aeruginosa through the hydrolysis of an enzyme substrate
Actively growing strains of Pseudomonas aeruginosa have an enzyme that cleaves the substrate in the reagent to produce blue fluorescence under ultraviolet light. The Pseudalert Test detects Pseudomonas aeruginosa at 1 cfu per 100 mL or 250 mL sample within 24 hours.
Pseudomonas aeruginosa cells rapidly grow and reproduce using the rich supply of amino acids, vitamins, and other nutrients present in the Pseudalert reagent.
ORDER
Expand All / Collapse All
Size Volume Supplier No. VWR Catalog Number Unit Price Quantity
200 tests per kit 100 mL 98-18078-00 35000-864 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty