Would you like to log in again?
Accessories
Supplier: VWR International
949VWCALUSB
ORDER
Expand All / Collapse All
Description Details VWR Catalog Number Unit Price Quantity
Calibration USB The calibration USB allows the user to calibrate the VWR Advanced Melting Point Apparatus to an external temperature device; The unit will use a biased offset for one of five specific temperature settings or Reference Samples; Reference samples are pure substrates with known melting point values 75840-102 Each Retrieving
Product and/or packaging contains recycled/renewable content
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFICATIONS
Environmentally Preferable Attribute Product and/or packaging contains recycled/renewable content
Environmental Certification RoHS
ACCESSORIES
Expand All / Collapse All
Accessories
Description Details VWR Catalog Number Unit Price Quantity
Capillary Tubes Capillary tubes with one end open for efficient sample collection; Tubes are 90mm long with an OD of 2mm and an ID of 1mm; Case of 2000, 90mm capillary tubes, open on one end 75840-018 Case of 2,000 Retrieving
Cooling Blocks Cooling blocks are designed to rapidly cool the oven back to room temperature; The copper material absorbs the heat and draws it out of the oven to provide efficient transition between tests involving various different melting points 75840-020 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty