RNA Purification Kits, Omni International
Supplier: Omni International
26-011B 26-012B 26-010B
75849-290EA 410.88 USD
75849-290 75849-306 75849-292
RNA Purification Kits, Omni International
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Omni Bead Mill Nucleic Acid Purification Kits are designed for use with bead mill homogenizers or vortex mixers to extract DNA or RNA from bacterial cells, fungal cells, tissues, soil or fecal samples.
  • Pre-filled, bead beating tubes containing an optimized amount of lysing matrix
  • High-yield, nucleic acid extractions from challenging samples such as soil and tough tissues
  • Silica-based, spin-column capture columns
Omni Bead Mill Nucleic Acid Purification Kits are designed for use with bead mill homogenizers or vortex mixers to extract DNA or RNA from bacterial cells, fungal cells, tissues, soil or fecal samples
Isolated RNA is suitable for downstream applications such as RT-PCR.
All Omni nucleic acid purification kits contain pre-filled bead beating tubes containing an optimized amount of lysing matrix to provide efficient and high yield nucleic acid extractions from challenging samples such as soil and tough tissues. Kits contain silica based spin-column capture columns, pre-filled bead beating tubes and nontoxic reagents.
Certifications: Pre-filled bead tubes are certified nuclease free.