Anti-IgG Rabbits Polyclonal Antibody

Supplier: MP Biomedicals
08643621
IC643621EA 226.46 USD
IC643621
Anti-IgG Rabbits Polyclonal Antibody
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbits
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR