Anti-6HIS Chicken Polyclonal Antibody

Supplier: MP Biomedicals
0859269
IC59269EA 330.58 USD
IC59269
Anti-6HIS Chicken Polyclonal Antibody
Antibodies
Type: Primary
Antigen: 6HIS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR