Anti-hCL(II) Mouse Monoclonal Antibody [clone: II-4C11]

Supplier: MP Biomedicals
08631713
IC631713EA 799.26 USD
IC631713
Anti-hCL(II) Mouse Monoclonal Antibody [clone: II-4C11]
Antibodies
Type: Primary
Antigen: hCL(II)
Clonality: Monoclonal
Clone: II-4C11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1/k
Reactivity: Human;Rabbit;Bovine;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR