You Searched For: 2-Thiazolylzinc bromide
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Catalog Number: (AAH58497-AD)
Supplier: Thermo Scientific Chemicals
Description: 2-Thiazolylzinc bromide, 0.5M in THF, CAS number: 173382-28-0, Molecular Formula: C3H2BrNSZn, Color: yellow to brown to black, Form: liquid, packaged under Argon in resealable ChemSeal bottles, Size: 50ML