    Sort by:
  • Search Within Results
Ristocetina
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Supplier:  MP Biomedicals
Description:   Soluble in 1 N HCl (50 mg/mL - clear and colorless to pale yellow); soluble in water (clear 5 mg+ 0.25 mL, amber solution).