    Sort by:
  • Search Within Results
1-Chloro-N,N-2-trimethylpropenylamine
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Acros Organics
Description:   1-Chloro-N,N,2-trimethylpropenylamine, Purity: 98.5+%, Cas no: 26189-59-3, MW: 133.62, MF: C6 H12 Cl N, form: Liquid, Color: Light yellow, Synonym: (1-Chloro-2-Methylpropenyl)Dimethylamine, TMCE, Tetramethyl-Alpha-Chloroenamine, Size: 25ML